Eosin Y (yellowish) 0.5% in aqueous solution for microscopy, Sigma-Aldrich®

Supplier: MilliporeSigma

1.09844.2500 1.09844.1000 1.09844.2500
EM1.09844.1000CS 912.92 CAD
EM1.09844.1000 EM1.09844.2500 CA1.09844.1000 CA1.09844.2500
Eosin Y (yellowish) 0.5% in aqueous solution for microscopy, Sigma-Aldrich®
Eosin Y (yellowish)
Formula: C₂₀H₆Br₄Na₂O₅
MDL Number: MFCD00005040
CAS Number: 17372-87-1

Order Now

Specification Test Results

Density 1.00 g/cm3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR