Spectroquant® NOVA 30A, Photometer
Supplier: MilliporeSigma
1.09748.0001 1.09748.0001
CA1.09748.0001EA 6643.57 CAD
CA1.09748.0001 EM1.09748.0001
Spectroquant® NOVA 30A, Photometer
Spectrophotometers and Photometers UV/Visible Spectrophotometers
The Spectroquant® NOVA 30A is capable of running all the important cell tests for wastewater analysis.
  • Barcode reading of all Spectroquant® tests: Automatic selection of methods with presentation of the results
  • Integrated QA/QC checks: Three modes, checking instrument, method and sample volume
  • Turbidity correction
Suitable for some critical tests, measurement to ppb levels, such as cyanide.