Zinc, powder, EMSURE® < 45 µm for analysis, Supelco®

Supplier: MilliporeSigma
Warning

1.08789.0500 1.08789.1000 1.08789.0500 1.08789.1000
EM1.08789.0500CS 10663.42 CAD
EM1.08789.0500 EM1.08789.1000 CA1.08789.0500 CA1.08789.1000
Zinc, powder, EMSURE® < 45 µm for analysis, Supelco®
Zinc
Formula: Zn
MDL Number: MFCD00011291
CAS Number: 7440-66-6
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Solubility (20 °C) insoluble,(reaction)
Melting Point 420 °C
Molar Mass 65.39 g/mol
Bulk Density 1800 - 2700 kg/m3
Boiling Point 908 °C (1013 hPa)
Vapor Pressure 1.33 hPa (487 °C)
Density 7.14 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 460 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR