Zinc, granular, EMSURE® ∼ 3-8 mm for analysis, Supelco®

Supplier: MilliporeSigma
Warning

1.08780.0500 1.08780.1000 1.08780.0500 1.08780.1000
EM1.08780.0500CS 16452.54 CAD
EM1.08780.0500 EM1.08780.1000 CA1.08780.0500 CA1.08780.1000
Zinc, granular, EMSURE® ∼ 3-8 mm for analysis, Supelco®
Zinc
Formula: Zn
MDL Number: MFCD00011291
CAS Number: 7440-66-6
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 419 °C
Molar Mass 65.39 g/mol
Boiling Point 906 °C (1013 hPa)
Density 7.13 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR