Zinc ≥99.9%, coarse powder, EMSURE® Reag. Ph. Eur. ∼ 0.3- 1.5 mm (14-50 mesh ASTM) for analysis, suitable for filling of reductors, Supelco®
Supplier: MilliporeSigma
|
Warning
|
1.08756.0250
1.08756.1000
1.08756.0250
1.08756.1000
EM1.08756.0250CS
41342.28
CAD
EM1.08756.0250
EM1.08756.1000
CA1.08756.0250
CA1.08756.1000
Zinc ≥99.9%, coarse powder, EMSURE® Reag. Ph. Eur. ∼ 0.3- 1.5 mm (14-50 mesh ASTM) for analysis, suitable for filling of reductors, Supelco®
Zinc
Formula:
Zn Boiling Pt: 907 °C (1013 hPa) Melting Pt: 419 °C Density: 7.13 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011291 CAS Number: 7440-66-6 EINECS: 231-175-3 UN: 3077 ADR: 9,III |
Specification Test Results
Assay (complexometric) | ≥ 99.9 % |
As (Arsenic) | ≤ 0.00001 % |
Cd (Cadmium) | ≤ 0.002 % |
Cu (Copper) | ≤ 0.001 % |
Fe (Iron) | ≤ 0.02 % |
Pb (Lead) | ≤ 0.005 % |
Sn (Tin) | ≤ 0.001 % |
Corresponds to Reag. Ph Eur |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...