Antimony(III) chloride, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Antimony trichloride

1.07838.0250 1.07838.1000 1.07838.0250 1.07838.1000
EM1.07838.0250CS 3694.04 CAD
EM1.07838.0250 EM1.07838.1000 CA1.07838.0250 CA1.07838.1000
Antimony(III) chloride, EMSURE® for analysis, Supelco®
Antimony (III) chloride
Formula: SbCl₃
MW: 228.12 g/mol
Boiling Pt: 223.5
Melting Pt: 73
MDL Number: MFCD00011212
CAS Number: 10025-91-9
UN: 1733
ADR: 8 II
Merck Index: 13,00713

Order Now

Specification Test Results

Solubility 931 g/l (20 °C) Hydrolysis
Melting Point 73 °C
Molar Mass 228.1 g/mol
Bulk Density 1600 kg/m3
Boiling Point 233 °C (1013 hPa)
Vapor Pressure 0.16 hPa (20 °C)
Density 3.14 g/cm3 (20 °C)
pH (H2O) acidic


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR