Antimony(III) oxide, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Antimony trioxide, Diantimony trioxide, di-Antimony trioxide

1.07836.0100 1.07836.1000 1.07836.0100 1.07836.1000
EM1.07836.0100CS 17964.6 CAD
EM1.07836.0100 EM1.07836.1000 CA1.07836.0100 CA1.07836.1000
Antimony(III) oxide, EMSURE® for analysis, Supelco®
Antimony (III) oxide
Formula: O₃Sb₂
MW: 291.52 g/mol
Boiling Pt: 1425
Melting Pt: 654
MDL Number: MFCD00011214
CAS Number: 1309-64-4
UN: 1549
Merck Index: 13,00717

Order Now

Specification Test Results

Solubility 2.70 mg/l (20 °C)
Melting Point 656 °C (sublimed)
Molar Mass 291.49 g/mol
Bulk Density 800 - 1300 kg/m3
Boiling Point 1550 °C (1013 hPa)
Vapor Pressure 13.3 hPa (660 °C)
Density 5.4 g/cm3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR