Antimony(III) oxide ≥99.0% (by bromometric analysis), Extra Pure, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Antimony trioxide, Diantimony trioxide, di-Antimony trioxide

1.07835.2500 1.07835.9050 1.07835.2500 1.07835.9050
EM1.07835.2500CS 6842.98 CAD
EM1.07835.2500 EM1.07835.9050 CA1.07835.2500 CA1.07835.9050
Antimony(III) oxide ≥99.0% (by bromometric analysis), Extra Pure, Supelco®
Antimony (III) oxide
Formula: O₃Sb₂
MW: 291.52 g/mol
Boiling Pt: 1550 °C (1013 hPa)
Melting Pt: 656 °C (subl.)
Density: 5.4 g/cm3
Storage Temperature: Ambient
MDL Number: MFCD00011214
CAS Number: 1309-64-4
UN: 1549
ADR: 6.1,III

Order Now

Specification Test Results

Solubility 2.70 mg/l (20 °C)
Melting Point 656 °C (sublimed)
Molar Mass 291.49 g/mol
Bulk Density 800 - 1300 kg/m3
Boiling Point 1550 °C (1013 hPa)
Vapor Pressure 13.3 hPa (660 °C)
Density 5.4 g/cm3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR