Selenium, black ≥99%, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.07714.0050 1.07714.0250 1.07714.1000 1.07714.0050 1.07714.0250 1.07714.1000
EM1.07714.0050CS 27403.52 CAD
EM1.07714.0050 EM1.07714.0250 EM1.07714.1000 CA1.07714.0050 CA1.07714.0250 CA1.07714.1000
Selenium, black ≥99%, EMSURE® for analysis, Supelco®
Selenium
Formula: Se
MDL Number: MFCD00134090
CAS Number: 7782-49-2
UN: 3288
ADR: 6.1,III

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Melting Point 217 °C
Molar Mass 78.96 g/mol
Bulk Density 1270 kg/m3
Boiling Point 685 °C (1013 hPa)
Vapor Pressure 0.001 hPa (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR