Phenylhydrazinium chloride, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.07253.0100 1.07253.0250 1.07253.0100 1.07253.0250
EM1.07253.0100CS 5512.62 CAD
EM1.07253.0100 EM1.07253.0250 CA1.07253.0100 CA1.07253.0250
Phenylhydrazinium chloride, EMSURE® for analysis, Supelco®
Phenylhydrazinium chloride
Formula: C₆H₉ClN₂
MDL Number: MFCD00012924
CAS Number: 59-88-1
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e16d43c3611e20636f31c95d3974860d83c8913f&vid=c26f549d3aed1b69060ac1042fd1114ca4edb2cb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 50 g/l (20 °C)
Melting Point 242 - 246 °C (decomposition)
Molar Mass 144.6 g/mol
Bulk Density 330 kg/m3
pH 2.6 - 2.9 (50 g/l, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR