2-Propanol ≥99.5% (by GC), EMPARTA® ACS for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.07022.2511 1.07022.4000 1.07022.9026 1.07022.2511 1.07022.4000 1.07022.9026
EM1.07022.2511CS 894.28 CAD
EM1.07022.2511 EM1.07022.4000 EM1.07022.9026 CA1.07022.2511 CA1.07022.4000 CA1.07022.9026
2-Propanol ≥99.5% (by GC), EMPARTA® ACS for analysis, Supelco®
2-Propanol
Formula: (CH₃)₂CHOH
Boiling Pt: 82 °C (lit.)
Melting Pt: –89.5 °C (lit.)
Density: 0.785 g/ml (25°C) (lit.)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
UN: 1219
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -89.5 °C
Molar Mass 60.1 g/mol
Boiling Point 82.4 °C (1013 hPa)
Vapor Pressure 43 hPa (20 °C)
Flash Point 12 °C
Refractive Index 1.378
Explosion Limit 2 - 13.4 %(V)
Density 0.786 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral
Ignition Point 425 °C
Evaporation Number 11


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR