Dichloromethane, EMPARTA® ACS analysis_for, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Methylene chloride

1.07020.2500 1.07020.4000 1.07020.6010 1.07020.2500 1.07020.4000 1.07020.6010
EM1.07020.2500CS 812.12 CAD
EM1.07020.2500 EM1.07020.4000 EM1.07020.6010 CA1.07020.2500 CA1.07020.4000 CA1.07020.6010
Dichloromethane, EMPARTA® ACS analysis_for, Supelco®
Dichloromethane
Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8
Melting Pt: -95
MDL Number: MFCD00000881
CAS Number: 75-09-2
UN: 1593
ADR: 6.1 III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 20 g/l (20 °C)
Melting Point -95 °C
Molar Mass 84.93 g/mol
Boiling Point 40 °C (1013 hPa)
Vapor Pressure 475 hPa (20 °C)
Refractive Index 1.42
Explosion Limit 13 - 22 %(V)
Density 1.33 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral
Ignition Point 605 °C DIN 51794
Evaporation Number 1.9


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR