Lithium hydroxide 98, Supelco®

Supplier: MilliporeSigma
Danger

1.05691.0100 1.05691.1000 1.05691.0100 1.05691.1000
EM1.05691.0100CS 79824.76 CAD
EM1.05691.0100 EM1.05691.1000 CA1.05691.0100 CA1.05691.1000
Lithium hydroxide 98, Supelco®
Lithium hydroxide
Formula: LiOH
MW: 23.95 g/mol
Boiling Pt: 924
Melting Pt: 462
MDL Number: MFCD00011095
CAS Number: 1310-65-2
UN: 2680
ADR: 8 II
Merck Index: 13,05556

Order Now

Specification Test Results

Solubility 71 g/l (20 °C)
Melting Point 462 °C
Molar Mass 23.95 g/mol
Bulk Density 550 kg/m3
Density 2.54 g/cm3
pH 12 (50 g/l, H2O, 50 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR