Hematoxylin, crystals for microscopy (C.I. 75290), Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: DcH-48, Hydroxybrazilin, (6aS,11bR)-6,6a,7,11b-Tetrahydroindeno[2,1-c]chromene-3,4,6a,9,10-pentaol, NSC 2785, Natural Black I, Hydroxybrasilin, (+)-Hematoxylin

M-Clarity™ quality level = MQ400

1.04302.0025 1.04302.0100 1.04302.0025 1.04302.0100
EM1.04302.0025CS 17266.24 CAD
EM1.04302.0025 EM1.04302.0100 CA1.04302.0025 CA1.04302.0100
Hematoxylin, crystals for microscopy (C.I. 75290), Sigma-Aldrich®
Hematoxylin
Formula: C₁₆H₁₄O₆
MW: 302.28 g/mol
MDL Number: MFCD00078111
CAS Number: 517-28-2

Order Now

Specification Test Results

Identity (UV/VIS-Spectrum) passes test
Transition range pH 5.0 - 7.2, yellow - violet
Water (according to Karl Fischer) ≤ 10 %
Suitability for microscopy passes test
Papanicolaous stain (cyanophilic Cytoplasm) blue-green to green
Papanicolaous stain (eosinophilic Cytoplasm) pink
Papanicolaous stain (Erythrocytes) red
Papanicolaous stain (Cell nuclei) blue to dark violet
Papanicolaous stain (Microorganisms) grey-blue
H&E staining (Cell nuclei) dark blue to dark violet
H&E staining (Cytoplasma) pink to red
H&E staining (intercellular substances) pink to red
H&E staining (Erythrocytes) yellow to orange


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR