Glutaraldehyde 25% in aqueous solution for electron microscopy, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

1.04239.1000 1.04239.2500 1.04239.0250 1.04239.1000 1.04239.2500
EM1.04239.0250CS 1045.04 CAD
EM1.04239.0250 EM1.04239.1000 EM1.04239.2500 CA1.04239.0250 CA1.04239.1000 CA1.04239.2500
Glutaraldehyde 25% in aqueous solution for electron microscopy, Sigma-Aldrich®
Glutaraldehyde
Formula: C₅H₈O₂
MDL Number: MFCD00007025
CAS Number: 111-30-8
UN: 1760
ADR: 8,II

Order Now

Specification Test Results

Assay (alkalimetric) 23.5 - 26.5 %
pH-value ≥ 3.0
Density (d 20 °C/ 4 °C) 1.060 - 1.065
Absorbance A (235 nm; 1:200 deluted with water; 1 cm) ≤ 0.3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR