Formaldehyde ≥37% aqueous solution Stabilised, GR Reag. Ph. Eur., ACS zA, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
Stabilization:
10% MeOH
MQ400
1.04003.1000
1.04003.2500
1.04003.5000
1.04003.9025
1.04003.1000
1.04003.2500
1.04003.9025
EM1.04003.1000CS
793.16
CAD
EM1.04003.1000
EM1.04003.2500
EM1.04003.5000
EM1.04003.9025
CA1.04003.1000
CA1.04003.2500
CA1.04003.5000
CA1.04003.9025
Formaldehyde ≥37% aqueous solution Stabilised, GR Reag. Ph. Eur., ACS zA, Sigma-Aldrich®
Formaldehyde
Formula:
CH₂O MW: 30.03 g/mol Boiling Pt: -19.3 Melting Pt: -92 Density: 1.09 g/cm3 (20 °C), 1.09 g/ml (25°C) (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00003274 CAS Number: 50-00-0 UN: 2209 ADR: 8 III Merck Index: 13,04259 |
Specification Test Results
Assay (acidimetric) | 36.5 - 38.0 % |
Assay (iodometric) | 36.5 - 38.0 % |
Identity | passes test |
Color | ≤ 10 Hazen |
Appearance of solution | passes test |
Free acid (as HCOOH) | ≤ 0.025 % |
Density (20 °C) | 1.080 - 1.090 g/ml |
Chloride (Cl) | ≤ 0.0001 % |
Sulfate (SO₄) | ≤ 0.002 % |
Heavy metals (as Pb) | ≤ 0.0002 % |
Fe (Iron) | ≤ 0.0001 % |
Methanol (GC) | 9.0 - 11.0 % |
Sulfated ash | ≤ 0.002 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...