Formaldehyde ≥37% aqueous solution Stabilised Histology acid and Calcium carbonate_free, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

Stabilization: 10% MeOH
MQ400

1.03999.1000 1.03999.2500 1.03999.9025 1.03999.1000 1.03999.2500 1.03999.9025
EM1.03999.1000CS 592.5 CAD
EM1.03999.1000 EM1.03999.2500 EM1.03999.9025 CA1.03999.1000 CA1.03999.2500 CA1.03999.9025
Formaldehyde ≥37% aqueous solution Stabilised Histology acid and Calcium carbonate_free, Sigma-Aldrich®
Formaldehyde
Formula: CH₂O
MW: 30.03 g/mol
Boiling Pt: -19.3
Melting Pt: -92
Density: 1.09 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
UN: 1198
ADR: 8 III
Merck Index: 13,04259

Order Now

Specification Test Results

Assay (acidimetric) 37.0 - 38.0 %
Heavy metals as Pb ≤ 0.0002 %
Fe (Iron) ≤ 0.0001 %
Methanol (GC) 8-12 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR