Iron, EMSURE® 10 µm for analysis, reduced, Supelco®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ300
1.03819.0100
1.03819.0500
1.03819.0100
1.03819.0500
EM1.03819.0100CS
10676.06
CAD
EM1.03819.0100
EM1.03819.0500
CA1.03819.0100
CA1.03819.0500
Iron, EMSURE® 10 µm for analysis, reduced, Supelco®
Iron
Formula:
Fe Boiling Pt: 3000 °C/1013 hPa Melting Pt: 1535 °C Density: 7.87 g/cm3 at 20 °C |
MDL Number:
MFCD00010999 CAS Number: 7439-89-6 UN: 3089 ADR: 4.1,III |
Specification Test Results
Assay (cerimetric) | ≥ 99.5 % |
Substances insoluble in sulfuric acid | ≤ 0.1 % |
Chloride (Cl) | ≤ 0.002 % |
Sulphide (S) | ≤ 0.002 % |
As (Arsenic) | ≤ 0.0002 % |
Pb (Lead) | ≤ 0.002 % |
Cu (Copper) | ≤ 0.002 % |
Mn (Manganese) | ≤ 0.002 % |
Zn (Zinc) | ≤ 0.002 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...