Silica gel sachet: 7 x 9 cm desiccant, with moisture indicator (orange gel), Supelco®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

1.03804.0001 1.03804.0001
EM1.03804.0001CS 2997.67 CAD
EM1.03804.0001 CA1.03804.0001
Silica gel sachet: 7 x 9 cm desiccant, with moisture indicator (orange gel), Supelco®
Silica gel
Formula: SiO₂
MW: 60.08 g/mol
Melting Pt: > 1000 °C
MDL Number: MFCD00011232
CAS Number: 63231-67-4

Order Now

Specification Test Results

Water absorption capacity (24 h, 80 % relative humidity) ≥ 20 %
Loss on drying (140°C, 3 h) ≤ 10 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR