Silica gel sachet: 4 x 7 cm desiccant, with moisture indicator_orange gel, Supelco®

Supplier: MilliporeSigma


MQ200

1.03803.0001 1.03803.0002 1.03803.0001 1.03803.0002
EM1.03803.0001CS 3878.9 CAD
EM1.03803.0001 EM1.03803.0002 CA1.03803.0001 CA1.03803.0002
Silica gel sachet: 4 x 7 cm desiccant, with moisture indicator_orange gel, Supelco®
Silica gel
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1710
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4

Order Now

Specification Test Results

Water absorption capacity (24 h, 80 % relative humidity) ≥ 20 %
Loss on drying (140°C, 3 h) ≤ 10 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR