N,N-Dimethylformamide ≥99.5% (by GC), EMPARTA®, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Formic acid dimethylamide, DMFA, Formyldimethylamine, N-Formyldimethylamine, Dimethylformamide, NSC 5356, DMF
1.03034.1000
1.03034.1011
1.03034.2500
1.03034.4000
1.03034.6025
1.03034.1000
1.03034.2500
1.03034.2511
1.03034.4000
1.03034.6025
EM1.03034.1000CS
2035.04
CAD
EM1.03034.1000
EM1.03034.1011
EM1.03034.2500
EM1.03034.2511
EM1.03034.4000
EM1.03034.6025
CA1.03034.1000
CA1.03034.1011
CA1.03034.2500
CA1.03034.2511
CA1.03034.4000
CA1.03034.6025
N,N-Dimethylformamide ≥99.5% (by GC), EMPARTA®, Supelco®
N,N-Dimethylformamide
Formula:
HCON(CH₃)₂ Boiling Pt: 153 °C (lit.) Melting Pt: –61 °C (lit.) Density: 0.944 g/ml (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00003284 CAS Number: 68-12-2 UN: 2265 ADR: 3,III |
|
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | -61 °C |
Molar Mass | 73.09 g/mol |
Boiling Point | 153 °C (1013 hPa) |
Vapor Pressure | 3.77 hPa (20 °C) |
Flash Point | 58 °C |
Refractive Index | 1.43 |
Explosion Limit | 2.2 - 16 %(V) |
Density | 0.94 g/cm3 (20 °C) |
pH | 7 (200 g/l, H2O, 20 °C) |
Ignition Point | 410 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...