Cesium nitrate 99, EMSURE® analysis_for, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Nitric acid cesium salt

1.02856.0025 1.02856.1000 1.02856.0025 1.02856.1000
EM1.02856.0025CS 18078.36 CAD
EM1.02856.0025 EM1.02856.1000 CA1.02856.0025 CA1.02856.1000
Cesium nitrate 99, EMSURE® analysis_for, Supelco®
Cesium nitrate
Formula: CsNO₃
MW: 194.91 g/mol
Boiling Pt: 737
Melting Pt: 414
Density: 3.68 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010963
CAS Number: 7789-18-6
EINECS: 232-146-8
UN: 1451
ADR: 5.1 III
Merck Index: 12,02057

Order Now

Specification Test Results

Assay (acidimetric after ion exchange) ≥ 99 %
Chloride (Cl) ≤ 0.002 %
Sulfate (SO₄) ≤ 0.005 %
Heavy metals (as Pb) ≤ 0.001 %
Fe (Iron) ≤ 0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR