Copper(II) oxide, fine wire ∼ 0.65 x 3 mm for elementary analysis (surface: CuO, core: Cu₂O), Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Cupric oxide, Copper oxide, Copper monoxide granulated

1.02765.0250 1.02765.1000 1.02765.0250 1.02765.1000
EM1.02765.0250CS 7427.58 CAD
EM1.02765.0250 EM1.02765.1000 CA1.02765.0250 CA1.02765.1000
Copper(II) oxide, fine wire ∼ 0.65 x 3 mm for elementary analysis (surface: CuO, core: Cu₂O), Supelco®
Copper (II) oxide
Formula: CuO
MW: 79.55 g/mol
Boiling Pt: 2000
Melting Pt: 1326
Density: 6.2 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010979
CAS Number: 1317-38-0
EINECS: 215-269-1
UN: 3077
ADR: 9 III
Merck Index: 13,02674

Order Now

Specification Test Results

Assay (oxygen demand as CuO) ≥ 55.0 %
Chloride (Cl) ≤ 0.005 %
Total carbon (C) ≤ 0.002 %
Total nitrogen (N) ≤ 0.002 %
Total sulfur (as SO₄) ≤ 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR