Copper(II) oxide, powder, EMSURE®, extra pure for analysis, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Cupric oxide, Copper oxide, Copper monoxide granulated

1.02761.0500 1.02761.9025 1.02761.0500 1.02761.9025
EM1.02761.0500CS 7869.98 CAD
EM1.02761.0500 EM1.02761.9025 CA1.02761.0500 CA1.02761.9025
Copper(II) oxide, powder, EMSURE®, extra pure for analysis, Supelco®
Copper (II) oxide
Formula: CuO
MW: 79.55 g/mol
Boiling Pt: 2000
Melting Pt: 1326 (dec.)
Density: 6.48 g/cm3 at 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00010979
CAS Number: 1317-38-0
UN: 3077
ADR: 9 III
Merck Index: 13,02674

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 1326 °C (decomposition)
Molar Mass 79.55 g/mol
Bulk Density 500 kg/m3
Density 6.48 g/cm3 (25 °C)
pH 7 (50 g/l, H2O, 20 °C) (slurry)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR