Copper, fine powder, EMSURE® < 63 µm for analysis, Supelco®

Supplier: MilliporeSigma

1.02703.0250 1.02703.1000 1.02703.0250 1.02703.1000
EM1.02703.0250CS 31363 CAD
EM1.02703.0250 EM1.02703.1000 CA1.02703.0250 CA1.02703.1000
Copper, fine powder, EMSURE® < 63 µm for analysis, Supelco®
Copper

EMD # 1.02703.1000 replaces and is identical to EMD # CX1925-4.

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
MDL Number: MFCD00010965
CAS Number: 7440-50-8
UN: 3089
ADR: 4.1 III
Merck Index: 13,02545

Order Now

Specification Test Results

Melting Point 1083 °C
Molar Mass 63.55 g/mol
Bulk Density 1290 kg/m3
Density 8.96 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR