Eosin Y (yellowish) 0.5% alcoholic solution for microscopy, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Tetrabromofluorescein disodium saltEosin Y disodium saltEosin 2252′,4′,5′,7′-Tetrabromofluorescein disodium saltSolvent Red 43Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoateDisodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate)Field's Stain B2,4,5,7-Tetrabromofluorescein disodium salt

1.02439.2500
CA1.02439.2500CS 1279.98 CAD
CA1.02439.2500
Eosin Y (yellowish) 0.5% alcoholic solution for microscopy, Sigma-Aldrich®
Eosin Y (yellowish)

Counterstain in H and E.


Eosin Y solution 0.5% alcoholic, is a ready-to-use solution that can be used for routine staining in the hematoxylin-eosin staining process (H and E) of sample material of human origin. The H and E staining process is the most frequently used staining method for histological material. Due to the hematoxylin solution, the nuclei will appear blue to dark violet in color and the Eosin Y solution will stain the cytoplasma and intercellular substances red-orange.Sufficient for approx. 1500 specimens/L. It is registered as IVD and CE product.

Formula: C₂₀H₆Br₄Na₂O₅
MW: 691.86 g/mol
Density: 0.8889 g/cm3 (20 °C)
Flash Pt: 19.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00005040
CAS Number: 17372-87-1
UN: 1170
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR