Calcium chloride, granular ∼ 0.5-2.0 mm for elementary analysis, fused, Supelco®

Supplier: MilliporeSigma
Warning

1.02083.0250 1.02083.1000 1.02083.0250 1.02083.1000
EM1.02083.0250CS 32549.58 CAD
EM1.02083.0250 EM1.02083.1000 CA1.02083.0250 CA1.02083.1000
Calcium chloride, granular ∼ 0.5-2.0 mm for elementary analysis, fused, Supelco®
Calcium chloride
Formula: CaCl₂
MDL Number: MFCD00010903
CAS Number: 10043-52-4

Order Now

Specification Test Results

Solubility 740 g/l (20 °C)
Melting Point 772 °C
Molar Mass 110.98 g/mol
Boiling Point 1600 °C
Density 2.15 g/cm3 (20 °C)
pH 8 - 10 (100 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR