Cadmium, coarse powder, EMSURE® ∼ 0.3-1.6 mm for analysis, suitable for filling of reductors, Supelco®

Supplier: MilliporeSigma
Danger

1.02001.0250 1.02001.0250 1.02001.1000
EM1.02001.0250CS 44021.96 CAD
EM1.02001.0250 CA1.02001.0250 CA1.02001.1000
Cadmium, coarse powder, EMSURE® ∼ 0.3-1.6 mm for analysis, suitable for filling of reductors, Supelco®
Cadmium

EMD # 1.02001.1000 replaces and is identical to EMD # 2001-4.

Formula: Cd
MW: 112.41 g/mol
Boiling Pt: 765
Melting Pt: 321
Density: 8.6 g/cm3 (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010914
CAS Number: 7440-43-9
EINECS: 231-152-8
UN: 3288
ADR: 6.1 III
Merck Index: 12,01649

Order Now

Specification Test Results

Cu (Copper) ≤ 0.002 %
Fe (Iron) ≤ 0.001 %
Pb (Lead) ≤ 0.01 %
Zn (Zinc) ≤ 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR