Bismuth(III) oxide ≥99.0% (by complexometric analysis), extra pure, Supelco®
Supplier: MilliporeSigma
Synonyms:
Bismuth Trioxide, Dibismuth trioxide
1.01862.9025
1.01862.9025
1.01862.1000
EM1.01862.9025EA
19800.56
CAD
EM1.01862.9025
CA1.01862.9025
EM1.01862.1000
Bismuth(III) oxide ≥99.0% (by complexometric analysis), extra pure, Supelco®
Bismuth (III) oxide
Formula:
BiO₃ MW: 465.96 g/mol Boiling Pt: 1890 Melting Pt: 825 Density: 8.93 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003462 CAS Number: 1304-76-3 EINECS: 215-134-7 Merck Index: 13,01273 |
Specification Test Results
Melting Point | 820 °C |
Molar Mass | 465.95 g/mol |
Bulk Density | 1000 kg/m3 |
Boiling Point | 1890 °C |
Density | 8.93 g/cm3 (20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...