Sodium hydroxide on support, granulated ∼ 0.8-1.6 mm elementary_analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Caustic soda

1.01567.1000 1.01567.0250 1.01567.1000
EM1.01567.0250CS 1781.97 CAD
EM1.01567.0250 EM1.01567.1000 CA1.01567.0250 CA1.01567.1000
Sodium hydroxide on support, granulated ∼ 0.8-1.6 mm elementary_analysis, Supelco®
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8 II
Merck Index: 14,08627

Order Now

Specification Test Results

Absorption capacity for CO₂ ≥ 50 %
Loss on ignition (800 °C) ≤ 25 %
Bulk density about 70 g/100 ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR