Aluminium, foil, EMSURE® for analysis, Width: 30 mm, Thickness: 0.3 mm, Supelco®

Supplier: MilliporeSigma

Synonyms: Aluminum

1.01057.0250 1.01057.1000 1.01057.1000 1.01057.0250
EM1.01057.0250CS 51204.64 CAD
EM1.01057.0250 EM1.01057.1000 CA10053-782 CA1.01057.0250
Aluminium, foil, EMSURE® for analysis, Width: 30 mm, Thickness: 0.3 mm, Supelco®
Aluminium
Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327
Melting Pt: 660
MDL Number: MFCD00134029
CAS Number: 7429-90-5
Merck Index: 13,00321

Order Now

Specification Test Results

Solubility (20 °C) (reaction)
Melting Point 660 °C
Molar Mass 26.98 g/mol
Boiling Point 2467 °C
Density 2.70 g/cm3 (20 °C)
Ignition Point 400 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR