Aluminium ≥90% stabilised, fine powder, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Aluminum

Stabilization: ∼ 2% Stabilised with fat

1.01056.0250 1.01056.1000 1.01056.0250 1.01056.1000
EM1.01056.0250CS 4939.08 CAD
EM1.01056.0250 EM1.01056.1000 CA1.01056.0250 CA1.01056.1000
Aluminium ≥90% stabilised, fine powder, Supelco®
Aluminium
Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2467 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.70 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
UN: 1396
ADR: 4.3 II
Merck Index: 13,00321

Order Now

Specification Test Results

Assay (complexometric) ≥ 90.0 %
Heavy metals (as Pb) ≤ 0.03 %
As (Arsenic) ≤ 0.0005 %
Fe (Iron) ≤ 1.0 %
Fat passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR