Vanadium(V) oxide, Extra Pure, Supelco®

Supplier: MilliporeSigma
Danger

1.00824.0250 1.00824.1000 1.00824.0250 1.00824.1000
EM1.00824.0250CS 29876.22 CAD
EM1.00824.0250 EM1.00824.1000 CA1.00824.0250 CA1.00824.1000
Vanadium(V) oxide, Extra Pure, Supelco®
Vanadium (V) oxide
Formula: O₅V₂
MDL Number: MFCD00011457
CAS Number: 1314-62-1
UN: 2862
ADR: 6.1,III

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Melting Point 690 °C
Molar Mass 181.88 g/mol
Bulk Density 400 - 600 kg/m3
Boiling Point 1750 °C (decomposition)
Density 3.36 g/cm3 (20 °C)
pH 4 (50 g/l, H2O, 20 °C) (slurry)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR