Titanium(IV) oxide ≥99.0% (by cerimetric analysis, on dry substance) [98.0-100.5% (cerimetric)], EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma

Synonyms: Titanic anhydride, Pigment White 6, Rutile, Titanium(IV) oxide, rutile, Titanium dioxide

1.00808.1000 1.00808.9025 1.00808.9050 1.00808.1000 1.00808.9025 1.00808.9050
EM1.00808.1000CS 2466.38 CAD
EM1.00808.1000 EM1.00808.9025 EM1.00808.9050 CA1.00808.1000 CA1.00808.9025 CA1.00808.9050
Titanium(IV) oxide ≥99.0% (by cerimetric analysis, on dry substance) [98.0-100.5% (cerimetric)], EMSURE® for analysis, Supelco®
Titanium (IV) oxide

In North America 1.00808.1000 replaces and has a comparable specification but a different pack size to TX0685-2

Formula: TiO₂
MW: 79.88 g/mol
Boiling Pt: 2900 °C (1013 hPa)
Melting Pt: 1855 °C
Storage Temperature: Ambient
MDL Number: MFCD00011269
CAS Number: 13463-67-7

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 1855 °C
Molar Mass 79.87 g/mol
Bulk Density 850 kg/m3
Boiling Point 2900 °C
pH 7 - 8 (100 g/l, H2O, 20 °C) (slurry)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR