BOD Cell Test Method: photometric 0.5 to 3000 mg/L Spectroquant®, MilliporeSigma
Supplier: MilliporeSigma
1.00687.0001 1.00688.0001 1.00687.0001 1.00688.0001
EM1.00687.0001CS 1008.04 CAD
EM1.00687.0001 EM1.00688.0001 CA1.00687.0001 CA1.00688.0001
BOD Cell Test Method: photometric 0.5 to 3000 mg/L Spectroquant®, MilliporeSigma
Environmental Test Kits
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times.
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*3 BOD nutrient salt mixture  1.00688 required