Formaldehyde solution 4% for histology, buffered pH 6.9, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

1.00496.5000 1.00496.9010 1.00496.5000 1.00496.9010
EM1.00496.5000CS 774.2 CAD
EM1.00496.5000 EM1.00496.9010 CA1.00496.5000 CA1.00496.9010
Formaldehyde solution 4% for histology, buffered pH 6.9, Sigma-Aldrich®
Formaldehyde

(= 10% Formaline solution)

Formula: CH₂O
MDL Number: MFCD00003274
CAS Number: 50-00-0

Order Now

Specification Test Results

Boiling Point 100 °C (1013 hPa)
Density 1.017 g/cm3 (20 °C)
pH 6.7 - 7.1 (H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR