Molybdenum(VI) oxide, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Molybdenum trioxide
1.00403.0100
1.00403.0500
1.00403.0100
1.00403.0500
EM1.00403.0100CS
50052.82
CAD
EM1.00403.0100
EM1.00403.0500
CA1.00403.0100
CA1.00403.0500
Molybdenum(VI) oxide, EMSURE® for analysis, Supelco®
Molybdenum (VI) oxide
Formula:
MoO₃ MW: 143.94 g/mol Boiling Pt: 1155 Melting Pt: 795 |
MDL Number:
MFCD00003469 CAS Number: 1313-27-5 UN: 3288 Merck Index: 13,06261 |
Specification Test Results
Solubility | 0.5 g/l (20 °C) |
Melting Point | 795 °C |
Molar Mass | 143.94 g/mol |
Bulk Density | 1200 kg/m3 |
Boiling Point | 1155 °C |
Density | 4.70 g/cm3 (26 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...