N,N-Dimethylformamide for peptide synthesis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: NSC 5356Formic acid dimethylamideDimethylformamideDMFADMFFormyldimethylamineN-Formyldimethylamine

1.00397.4000 1.00397.6025 1.00397.6025 1.00397.2500 1.00397.2500 1.00397.4000
EM1.00397.4000EA 1536.27 CAD
EM1.00397.6025 EM1.00397.4000 CA1.00397.2500 EM1.00397.2500 CA1.00397.4000 CA1.00397.6025
N,N-Dimethylformamide for peptide synthesis, Supelco®
N,N-Dimethylformamide
Formula: HCON(CH₃)₂
MW: 73.09 g/mol
Boiling Pt: 153 °C (1013 hPa)
Melting Pt: –61 °C
Density: 0.949 g/cm³ (20 °C)
Flash Pt: 57.5 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://ca.vwr.com/stibo/web/std.lang.all/25/06/2302506.jpg https://ca.vwr.com/stibo/web/std.lang.all/06/12/16640612.jpg

Order Now

Specification Test Results

Purity (GC) ≥ 99.9 %
Identity (IR) conforms
Color ≤ 10 Hazen
Acidity ≤ 0.0005 meq/g
Alkalinity ≤ 0.0002 meq/g
Amines (Dimethylamine) ≤ 10 ppm
Evaporation residue ≤ 10
Water ≤ 300 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR