Nunc® Lab-Tek™ II Chambered Coverglass, Thermo Scientific
Supplier: Thermo Fisher Scientific
155379 155360 155382 155409
CA-62407-052CS 2721.16 CAD
CA-62407-052 CA62407-050 CA62407-054 CA62407-056
Nunc® Lab-Tek™ II Chambered Coverglass, Thermo Scientific
Cover Glasses
Culture and view living cells on a single device
Thermo Scientific™ Nunc™ Lab-Tek™ II Chambered Coverglass includes a thin coverglass that provides the optimum optical characteristics required for high-magnification microscopy and confocal image analysis.