VWR® UV-3100PC, V-3000-PC Spectrophotometers, UV-Vis Scanning and Vis
Supplier: VWR
VWR®
UV-3100PC V-3000PC
10037-438CS 7742.42 CAD
10037-438 10037-440
VWR® UV-3100PC, V-3000-PC Spectrophotometers, UV-Vis Scanning and Vis
Spectrophotometers and Photometers UV/Visible Spectrophotometers
Two fully scanning spectrophotometers for advanced measurements and quality control.
  • Large LCD display (320×240 pixels)
  • Full scan mode
  • Easily accessible USB port for data import and export
  • Supplied application software 'UV-Vis Analyst' allows PC control of the spectrophotometer
  • Methods for PC control or standalone include: Basic mode, quantitative, wavelength scan, kinetics, multi-wavelength, DNA/protein
  • The range of accessories includes an automatic 4-cell changer and Peltier thermostatted single cell holder
Reliable, robust and easy to use instruments with a range of accessories that will support the creation of a personal spectrophotometer measuring station. The application software offers all the important methods expected from a 'universal’ instrument. All housed in a robust cover with a well designed optical bench.
Ordering information: Supplied with 4 optical glass cells, 2 quartz cells (excludes model V3000-PC), dust cover, USB cable, USB (8 GB) stick, and UV-Vis analyst software for PC control.