Pierce™ Bacterial Protein Extraction Reagents, Thermo Scientific
Supplier: Thermo Scientific
78243 78248 78260 78266 90078 90079 90082
CAPI78243EA 520.43 CAD
CAPI78243 CAPI78248 CAPI78260 CAPI78266 CAPI90078 CAPI90079 CAPI90082
Pierce™ Bacterial Protein Extraction Reagents, Thermo Scientific
Protein Extraction Protein Extraction Reagents
Thermo Scientific B-PER Bacterial Protein Extraction Reagents are designed to extract soluble protein from bacterial cells without harsh chemicals or mechanical disruption.
  • Lysis of bacteria occurs directly in cell culture media
  • No centrifugation steps
  • Compatible with the addition of protease inhibitors, and the resulting protein extract can be used in protein assays, typical affinity purification methods (eg. GST, 6XHis), and other applications
These easy-to-use cell lysis reagents are nonionic detergent solutions that effectively disrupt cells and solubilize native or recombinant proteins without denaturation.
Certain formulations include DNase I and lysozyme enzyme supplements to improve yield of large molecular weight proteins that are difficult to purify. B-PER Reagents are compatible with the addition of standard protease inhibitors and with downstream protein purification methods, such as GST purification and His-tagged immobilized metal affinity chromatography (IMAC).
Delivery information: Kit contains B-PER® Reagent, 250 mL; Lysozyme, 50 mg/mL, 0.5 mL; and DNase I, 2500 U/mL, 0.5 mL. Separate reagents are also available.