Cycloheximide, Sigma-Aldrich®

Supplier: MilliporeSigma

239763-1GM 239763-5GM
CA80510-794EA 282.73 CAD
CA80510-794 CA80510-796
Cycloheximide, Sigma-Aldrich®
Cycloheximide

Cycloheximide is an antifungal antibiotic that inhibits protein synthesis in eukaryotes but not prokaryotes


Competitively inhibits hFKBP12 (Ki = 3.4 µM). It triggers apoptosis in HL-60 cells, T cell hybridomas, Burkitt’s lymphoma cells, and other cell types. Soluble in ethanol or methanol.

Formula: C₁₅H₂₃NO₄
MDL Number: MFCD00082346
CAS Number: 66-81-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3abdf7ad3f7b3ea9e67fba3b6f3970c6627794be&vid=6f07d0130a35bc66b22ecee29fc6c31b20823099&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR