ClipTip® 384 Pipette Tips, 12.5 μl, Thermo Scientific
Supplier: Thermo Scientific
94410050 94410053 94420053
10030-796CS 1353.93 CAD
10030-796 10030-798 10030-800
ClipTip® 384 Pipette Tips, 12.5 μl, Thermo Scientific
Pipette Tips
Thermo Scientific ClipTip® 384 pipette tips feature a unique ‘Snap and Lip’ seal technology to attach and seal the tips on each channel with minimal tip attachment and ejection force.
  • Available in non-sterile, sterile, and filtered formats
  • Certified free of RNase, DNase, DNA, ATP, and pyrogen contamination
  • Each tip insert has ink-jetted lot and volume information
ClipTip® 384 model selection is optimized for 384-microplate and molecular biology applications. The tips are available in three sizes and packaging options, with 12.5, 30, and 125 μl tips. AMore Product Information