CritiCore Cleanroom Scrub Pants (Inner Wear)
Supplier: CritiCore Protective Wear
CritiCore
10032-422 10032-426 10032-414 10032-420 10032-412 10032-424 10032-416 10032-418
10032-424EA 70.22 CAD
10032-424 10032-412 10032-426 10032-414 10032-416 10032-418 10032-420 10032-422
CritiCore Cleanroom Scrub Pants (Inner Wear)
Workwear
These scrub pants provide an excellent base-layer of protection for any cleanroom garment system
  • Drawstring front/elastic back
  • Double needle felled seams
  • Color-coded sizing loop
  • Lint-free fabric
Manufactured with a durable, lightweight micro-denier continuous filament polyester material to elimate a source for lint. The material has a sandwash finish for soft-touch comfort and moisture wicking properties.
They feature a combination elastic back and drawstring front for secure comfort, and the leg opening has an elastic back with a three-position snap for an adjustable closure.
Ordering information: Available in Navy only. Custom order product with extended lead time. Contact your Avantor sales representative for more information.