SP Bel-Art 'Space Saver' Non-Vacuum Desiccators, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F42030-0000 F42031-0000 F42032-0000
24988-199CS 484.81 CAD
24988-199 70690-134 70690-136
SP Bel-Art 'Space Saver' Non-Vacuum Desiccators, Bel-Art Products, a part of SP
Desiccators Desiccators, Plastic
Maximize your space in the laboratory or classroom.
  • Small benchtop footprint
  • Shatterproof
  • Clear polycarbonate top
  • Heavy-walled design
  • Neoprene O-ring assures a tight seal
In less than 12" of benchtop space, these strong, shatterproof desiccators will provide dry storage and protection for moisture-sensitive products
All models have extra heavy walls and include a 0.32 cm (⅛") thick perforated plate. Disposable or reusable desiccant can be placed on or under this plate. A neoprene O-ring assures a tight seal down to 0˚C without the need for grease (More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable