Samco™ Special Purpose Transfer Pipettes, Thermo Scientific
Supplier: Thermo Fisher Scientific
292 293 295 292
14670-430CS 1212.42 CAD
14670-430 14670-433 14670-439 490014-724
Samco™ Special Purpose Transfer Pipettes, Thermo Scientific
Pipettes Pasteur/Transfer Pipettes
Samco™ Special Purpose Transfer Pipettes are ideal for delivering drops onto slides.
Made of pliable, nontoxic polyethylene, the low-affinity surface reduces loss of cells and valuable proteins due to binding.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO14001 (Certificate Registration No. EMS 752603)