Bead Ruptor Pre-Filled Bead Tubes, 30 ml, Omni International
Supplier: Omni International
19-6350
10032-522PK 545.76 CAD
10032-522
Bead Ruptor Pre-Filled Bead Tubes, 30 ml, Omni International
Tubes Centrifuge Tubes
Bead beating is the most effective and verstile lysis method available.
  • Non sterile
30 ml polypropylene tube with leakproof screw cap. Vacuum chamber tested to 95 kPa. Pre-filled with lysing matrix. For use on the Omni International Bead Ruptor 24. Compatible with solvents, alcohols, chlorinated and aromatic hydrocarbons (except xylene), and ketones.
Omni International offers users the ultimate flexibility with a range of bead beating tubes and lysing matrices to support efficient homogenization of a wide range of samples. Bead Ruptor bead beating tubes are pre-filled with an optimized amount of lysing matrix to provide efficient, consistent, and high-yield sample homogenates.
Ordering information: Unit includes 50 bead beating tubes pre-filled with lysing matrix.