Amplitube™ PCR Reaction Strips, Simport Scientific
Supplier: Simport Scientific
T321-1B T321-1N T321-1G T321-1R T321-1Y T320-1N T320-1B T320-1G T320-1R T320-1Y T321-1B T321-1G T321-1R T321-1Y T321-2N T321-2B T321-2G T321-2R T321-2Y T321-2N T321-2B T321-2G T321-2R T321-2Y T321-3N T320-10N T320-10B T320-10G T320-10R T320-10Y T321-1B
CA48648-502CS 58.4 CAD
CA48648-502 CA48648-503 CA48648-504 CA48648-505 CA48648-506 CA60872-639 CA60872-641 CA60872-644 CA60872-648 CA60872-650 89428-996 89428-998 89429-000 89429-002 89429-004 89429-006 89429-008 89429-010 89429-012 CA11028-424 CA11028-426 CA11028-428 CA11028-430 CA11028-432 89511-392 89511-394 89511-396 89511-398 89511-400 89511-402 490010-418
Amplitube™ PCR Reaction Strips, Simport Scientific
Tubes PCR Tubes
PCR Reaction strips include 8 or 12 integral 0.2 ml tubes with ultra-thin sidewalls and bottoms to ensure uniform and efficient temperature transfer.
  • Autoclavable
Non-attached cap strips are available in a dome or flat top design to ensure a perfect closure during the whole thermal cycle. Cap strips are not included and must be ordered separately.
All strips are molded of polypropylene under the most stringent conditions and are offered colorless, or in four different color varieties.
They are more easily handled than single tubes, and will precisely fit standard well spacing. They can also be used with 8- and 12-channel hand-held pipettors.
Certifications: Certified RNase-, DNase-, Pyrogen-, and DNA-free.
Packaging: Packed in tamper-proof resealable bags.