Amplitube™ PCR Reaction Tubes, 0.2 ml, Simport Scientific
Supplier: Simport Scientific
T325-1N T325-1B T325-1G T325-1R T325-1Y T325-2B T325-2N T325-2G T325-2R T325-2Y T325-2B T325-2G T325-2N T325-2R T325-2Y T325-2VB T325-2VG T325-2VR T325-2VN T325-2VY
CA48648-124CS 102.74 CAD
CA48648-124 CA48648-125 CA48648-126 CA48648-128 CA48648-129 CA48648-141 CA48648-142 CA48648-143 CA48648-144 CA48648-145 CA60872-628 89429-014 89429-016 89429-018 89429-020 89429-022 CA10030-906 CA10030-908 CA10030-910 CA10030-938 CA10030-940
Amplitube™ PCR Reaction Tubes, 0.2 ml, Simport Scientific
Tubes PCR Tubes
Amplitube™ PCR Reaction tubes are made of transparent superior grade polypropylene for improved viewing of the sample contents.
  • Attached hinged caps are either dome or flat-topped
  • Caps provide positive sealing during thermal cycling and prevent evaporation
  • Caps can be easily opened and closed with one hand
  • Autoclavable
Tubes are available with or without an integral shield that prevents contamination with surface of lid. A frosted writing surface is provided for sample identification.
Ultra-thin wall design ensures rapid thermal transfer andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable