Semi-Micro Vacuum Sublimation Apparatus, Ace Glass Incorporated
Supplier: Ace Glass
9546-04 9546-06 9546-08 9546-10
89075-108EA 186.87 CAD
89075-108 89075-110 89075-112 89075-114
Semi-Micro Vacuum Sublimation Apparatus, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
A semi-micro sublimer is used by a number of synthesis laboratories.
  • Handles a variety of crystal types
  • Large joint is a standard taper
  • Side take-off [ST] inner joint
Apparatus proportions have been found to be optimum for handling a variety of crystal types. The circulatory stem may be removed and collecting tube filled with dry ice, acetone. Complete unit consists of body, head and circulatory stem. Large joint is a standard taper 29/26, body length approximately 50 mm. Side take-off is a standard taper 14/20 inner joint. Outlet tube on 89075-108 has 8 mm O.D.; outlet tube on 89075-110 has 9 mm O.D.